Cole-Parmer Ultrasonic Four-Element Probe; 1/8" Dia
Representative image only.
Cole-Parmer Item # EW-04712-30
Maximize productivity by processing numerous samples simultaneously
  • Probes process from 250 μL to 10 mL sample volume
  • Energy delivered to each tip is uniform with 2% variation
  • 3 year warranty
PRODUCT OPTIONS
$1,645.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Sample Size (mL)10
  • DescriptionUltrasonic Four-Element Probe; 1/8" Dia
  • Warranty3 year
3 Year WarrantyConformite EuropeenneISO9001:2000 Certified Supplier
Key Features
  • Probes process from 250 μL to 10 mL sample volume
  • Energy delivered to each tip is uniform with 2% variation
  • 3 year warranty
More About this Item
The multi-element probes screw into the converter in place of the 1⁄2" (13 mm) probe. The energy delivered to each tip is uniform with 2% variation. The spacing between the tips is 2 2⁄32" (18 mm). Coupler and stepped microtips measure 10 1⁄4" (260 mm) long. Probes process from 250 μL to 10 mL sample volume and consist of aluminum coupling and 4-, 8-, or 24-element probes.