Cole-Parmer Ultrasonic Processor Sound-Abating enclosure; 30"H

Representative image only.
Cole-Parmer Item # EW-04710-48

$724.00USD / Each
Often Bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Height (in)30
  • Width (in)14
  • Depth (in)14
  • Height (cm)76.2
  • Width (cm)35.56
  • Depth (cm)35.56
  • DescriptionUltrasonic Processor Sound-Abating enclosure; 30"H

More About this Item

Permits extended processing by reducing noise to a comfortable level. Includes a support rod and converter clamp. Enclosure measures 14"W x 30"H x 14"D.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat