Cole-Parmer Valved Acrylic Flowmeter, 50 mm Scale for Air, 0.04-0.5 LPM
Representative image only.
Cole-Parmer Item # EW-32460-40
Meter with valve provides flow control through a highly durable meter body
  • Ideal for process plant applications on air sampling equipment, gas analyzers and chemical feed systems for water treatment
  • Integrated precision valve allows precise manual flow across the full scale
  • The flexible design allows for panel or bench mounting
  • 50-mm flowmeter
  • For air 0.04 to 0.5 LPM
PRODUCT OPTIONS
$89.00USD / Each
BUY MORE & SAVE
  • 1-3$89.00USD / Each
  • 4-9$80.10USD / Each
  • 10-19$75.65USD / Each
  • 20+$71.20USD / Each
$262.00USD / Each
for an additional
$262.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±5% full-scale
  • Repeatability±0.5% full-scale
  • Min Air Flow Rate (mL/min)40
  • Max Air Flow Rate (mL/min)500
  • Min Flow Rate (mL/min)40
  • Max Flow Rate (mL/min)500
  • Material FittingChrome-plated brass
  • Material ValveBrass
  • Material O-RingBuna-N
  • Material FloatGlass
  • Material HousingAcrylic
  • Max Operating Temperature (° F)150
  • Max Operating Temperature (° C)65
  • Gas TypeAir
  • Media TypeAir
  • Display Type50 mm Scale
  • DescriptionValved Acrylic Flowmeter, 50 mm Scale for Air, 0.04-0.5 LPM
Includes
Flowmeter
Key Features
  • Ideal for process plant applications on air sampling equipment, gas analyzers and chemical feed systems for water treatment
  • Integrated precision valve allows precise manual flow across the full scale
  • The flexible design allows for panel or bench mounting
  • 50-mm flowmeter
  • For air 0.04 to 0.5 LPM
More About this Item
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters’ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (sold separately) which allows for mobility from bench to bench.
Note: There are many additional types of acrylic flowmeters not listed here. Contact our Application Specialists for quotes on acrylic meters with special requirements.