Cole-Parmer VapLock™ Adapters for Exhaust Filters, Straight, Adjustable, Black Acetal, 1/4" NPT(M) x 1/4" NPT(F)
Representative image only.
Cole-Parmer Item # EW-12019-96
Various adapter configurations allow quick and easy connection to emission sources
  • Constructed of durable black acetal
PRODUCT OPTIONS
$42.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialAcetal
  • Connection TypeNPT(M) x NPT(F), adjustable
  • Thread size NPT(M) (port 2) (in)1/4
  • Thread size NPT(F) (port 1) (in)1/4
  • ColorBlack
  • DescriptionAdjustable Filter Adapter, 1/4\" NPT(F), black acetal; 1/ea
Key Features
  • Constructed of durable black acetal
More About this Item
VapLock exhaust filters have a 1/4" NPT(F) port and may require an adapter to work with your container. Adjustable adapters provide flexibility in positioning the filter to optimize use of space.