Cole-Parmer VapLock™ Fitting, Polypropylene, Straight, Hose Barb to Threaded Adapter, 1/16" ID x 1/8" NPT(M); 1/EA
Representative image only.
Cole-Parmer Item # EW-12026-35
Designed for secure leak-resistant connections
  • Constructed of durable polypropylene
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Connection TypeBarb x NPT(M)
  • Thread size NPT(M) (port 2) (in)1/8
  • Barb Size / Tubing Id (Port 1) (in)1/16
  • ColorGray
  • Description Fitting, Polypropylene, Straight, Hose Barb to Threaded Adapter, 1/16\" ID x 1/8\" NPT(M); 1/EA
Key Features
  • Constructed of durable polypropylene
More About this Item
Adapters for flexible tubing let you connect 1/16" to 3/4" ID soft-wall tubing to NPT(F) ports. Adapters are made of polypropylene. Wrap NPT threads 3 to 4 times with PTFE tape and wrench tighten for leak-resistant connections.