Cole-Parmer VapLock™ Luer Fitting, PVDF, Straight Bulkhead, Female Luer x 1/4-28 UNF(M) Bottom Sealing Thread; 1/EA
Representative image only.
Cole-Parmer Mfr # BSFTLL-J1A – Item # EW-12028-62
Quick connection and release make these luer fittings excellent for many applications
  • Great for connecting to syringes and small tubing
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyvinylidene fluoride (PVDF)
  • Number of Ports2
  • Connection Type-Port 1Straight female luer, bulkhead
  • Connection Type-Port 21/4-28 UNF(M) bottom sealing thread
  • DescriptionLuer Fitting, PVDF, Straight Bulkhead, Female Luer x 1/4-28 UNF(M) Bottom Sealing Thread; 1/EA
Key Features
  • Great for connecting to syringes and small tubing
More About this Item
Choose from fittings constructed from durable polypropylene (PP) or chemically inert PVDF.