Cole-Parmer VapLock™ Plug, PTFE, Luer Taper Plug; 1/Ea
Representative image only.
Cole-Parmer Item # EW-12027-52
Use plugs to seal unused ports for a leak-resistant system
  • Seal unused manifold and bottle cap ports
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPTFE
  • Connection TypeUNF(M)
  • Thread size UNF(M) (port 1) (in)Luer Taper Plug
  • ColorWhite
  • DescriptionPlug, PTFE, Luer Taper Plug; 1/Ea
Key Features
  • Seal unused manifold and bottle cap ports
More About this Item
Bottom-sealing UNF thread options include 1/4-28 and 5/16-24, and do not require PTFE tape for leak-resistant connections.