Cole-Parmer Versatile Cap Silicone O-Ring, 53B; 5/Pk
Representative image only.
Cole-Parmer Item # EW-06061-32
Replacement o-rings for Versatile Carboy Caps and Adapter Inserts
  • High purity platinum-cured silicone
SPECS & DESCRIPTION
Specifications & Description
  • Cap MaterialPlatinum-cured silicone
  • Cap / Stopper Size53B
  • AutoclavableYes
  • DescriptionVersatile Cap Silicone O-Ring, 53B; 5/Pk
Key Features
  • High purity platinum-cured silicone
More About this Item
Autoclavable.