Cole-Parmer Viscosity Cup #5, 460 to 1725 cSt, with NIST-Traceable Report
Representative image only.
Cole-Parmer Item # EW-08700-69
Simple and convenient – no complicated training needed
PRODUCT OPTIONS
$305.10USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Viscosity460 to 1725 centistokes
  • Cup Number5
  • Nist-TraceableYes
  • DescriptionViscosity Cup #5, 460 to 1725 cSt, w/NIST-traceable report
More About this Item
Measure the viscosity of liquids such as paint, lacquer, oil, and syrups. Simply insert the cup into your sample and lift the viscosity cup out, starting a stopwatch/timer. Cup is made of series 400 stainless steel and requires 44 mL of sample for the test. After withdrawing the cup, measure the time it takes until the first break in the liquid stream. A conversion sheet is supplied that will convert the Zahn time to a centistokes value.
Cup includes an NIST-traceable calibration report supplied by the manufacturer to meet your quality program requirements.