Cole-Parmer Waste System Cap w/ Exhaust Filter, 4 x 1/8" & 3 x 1/4" OD, 1 x 1/4" or 3/8" barb
Representative image only.
Cole-Parmer Item # EW-06063-24
Protect your lab from harmful solvent vapors
  • Carbon exhaust filter traps fumes
  • Tubing attaches securely to prevent escaping vapors and spills
  • Elbow adapter included to save on vertical space
SPECS & DESCRIPTION
Specifications & Description
  • Cap MaterialPolypropylene
  • Cap / Stopper Size83 mm
  • Cap ColorWhite
  • Special Features4 x 1/8\" & 3 x 1/4\" OD tubing, 1 x 1/4\" or 3/8\" hose barb
  • DescriptionWaste System Cap w/ Exhaust Filter, 4 x 1/8\" & 3 x 1/4\" OD, 1 x 1/4\" or 3/8\" barb
Key Features
  • Carbon exhaust filter traps fumes
  • Tubing attaches securely to prevent escaping vapors and spills
  • Elbow adapter included to save on vertical space
More About this Item
These carboy cap assemblies feature tubing ports with fittings to ensure a leak-tight connection and an activated carbon exhaust filter to remove solvent fumes from escaping air. Ideally suited for HPLC waste collection, the cap is constructed of polypropylene and includes a replaceable sulfur-cured EPDM gasket. Tubing fittings are made of acetal and ETFE for compatibility with HPLC-grade solvents. Caps fit standard carboys and jerricans with 83-mm diameter neck.