Cole-Parmer Waste System, UN/DOT, 70mm Cap, 4x1/8" & 3x1/4" OD Tubing, 1x1/4"-3/8" Hose Barb; 20L

Representative image only.
Cole-Parmer Item # EW-06062-53
Provide safe HPLC waste disposal in your lab
  • Certified for UN/DOT Packaging Group II for hazardous chemicals
  • Carbon filter captures potentially harmful vapors
  • Multiple ports adapt cap to any HPLC system
  • Filter adapter elbow provides storage flexibility, especially for tight lab spaces

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (L)20
  • Capacity (Gal)5.28
  • MaterialHigh Density Polyethyene (HDPE)
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size70 mm
  • Cap ColorWhite
  • Bottle ColorWhite
  • Special Features4 x 1/8\" and 3 x 1/4\" OD Tubing
  • AutoclavableNo
  • Tubing ID1 x 1/4\" to 3/8\" Hose Barb
  • DescriptionWaste System, UN/DOT, 70mm Cap, 4x1/8\" & 3x1/4\" OD Tubing, 1x1/4\"-3/8\" Hose Barb; 20L

Key Features

  • Certified for UN/DOT Packaging Group II for hazardous chemicals
  • Carbon filter captures potentially harmful vapors
  • Multiple ports adapt cap to any HPLC system
  • Filter adapter elbow provides storage flexibility, especially for tight lab spaces

More About this Item

This solvent waste system allows easy and safe disposal of HPLC waste in the lab. The system includes a UN/DOT container with two caps; one with ports for waste collection and one to seal for transportation. Caps come with a variety of port sizes to be used with complex HPLC systems. When not in use, the ports can be plugged.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat