Cole-Parmer Water, HPLC Grade, 2.5 L

Representative image only.
Cole-Parmer Item # EW-88401-24
Quality and consistency your lab can count on
  • Water meets HPLC grade and ACS reagent grade standards
  • Suitable for liquid chromatography
  • Water comes in 2.5 L size and packaged in an clear glass bottle

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeHPLC
  • Empirical FormulaH2O
  • FormLiquid
  • Synonymsdihydrogen oxide, dihydrogen monoxide
  • Volume (mL)2500
  • Formula Weight18.02
  • PackagingClear Glass Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7732-18-5
  • Shelf Life (months)18
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • DescriptionWater, HPLC Grade, 2.5 L

Key Features

  • Water meets HPLC grade and ACS reagent grade standards
  • Suitable for liquid chromatography
  • Water comes in 2.5 L size and packaged in an clear glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat