Cole-Parmer White Light Replacement Tube for UV Viewing Cabinets, 25-Watts
Representative image only.
Cole-Parmer Item # EW-97621-50
Get uniform UV illumination for optimal viewing
  • Stay comfortable with soft rubber viewport viewing area
  • Removable bottom panel allows cabinet to be placed over large samples or a transilluminator
  • Block out ambient light with lightweight access curtain
  • Protect your eyes with contrast control filter
PRODUCT OPTIONS
$31.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Power (watts)25
  • DescriptionWhite Light Replacement Tube for UV Viewing Cabinets, 25-Watts
Key Features
  • Stay comfortable with soft rubber viewport viewing area
  • Removable bottom panel allows cabinet to be placed over large samples or a transilluminator
  • Block out ambient light with lightweight access curtain
  • Protect your eyes with contrast control filter
More About this Item
The customizable viewing cabinets are designed to accommodate one or two 8-watt lamps (sold separately). Lamps plug directly into the cabinet housing as needed and can be removed for handheld use. Using two lamps provides uniform illumination from two sides of the cabinet. Cabinet features interior overhead white light, included with unit, for basic sample illumination.
The high-performance viewing cabinets feature high-intensity 15-watt UV tubes. Control switches, for selecting UV or white light, are conveniently located on the top of cabinet. Models 97621-10 and -15 feature two each of shortwave and longwave UV tubes mounted on one side of the cabinet. Models 97621-20 and -25 have four longwave UV tubes mounted on one side. Models 97621-30 and -35 have two shortwave and two longwave tubes mounted on two sides.
Product may contain trace amounts of mercury; cautions should be used in breakage, cleanup, shipping and/or disposal.