International Products Corp Zymit® Low-Foam Enzyme Cleaner; 210 kg

Representative image only.
International Products Corp Mfr # Z-9790 – Item # EW-18100-16
Ideal for use in ultrasonic cleaners and glassware washers
  • Available in 1-L bottles to 210 kg plastic drums!

SPECS & DESCRIPTION

Specifications & Description

  • FormatLiquid
  • Dilution1:100 to 2:100, pH 7.5
  • ApplicationsBlood, Emulsifiers, Fingerprints, Fragrances, Gels, Proteins, Starch
  • DescriptionZymit® Low-Foam Enzyme Cleaner; 210 kg

Key Features

  • Available in 1-L bottles to 210 kg plastic drums!

More About this Item

This enzyme-based liquid cleaner removes biological materials from chemical, food, and pharmaceutical equipment. Cleaner has pH of 7.5 allowing you to use it with delicate materials and metals. Solution is USDA approved.

Zymit Low-foam is a liquid dual-enzyme and detergent formula for removal of protein and starch based soils.

Zymit Low-foam is used to clean diagnostic and precision instruments, labware, photographic film equipment, and daycare furnishings and many other surfaces. Target soils include food, grass, fat, sweat, mucous, tissue, feces and sebum. Solution is safe for workers and the environment - contains no phosphates, CFCs, ODCs, phenols or borates.

Designed for use in ultrasonic baths, automatic washers and high-agitation applications. A 1% to 2% solution is effective in most applications. Typical pH: 7.5. NSF-registered for USDA A1 use.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat