Fitting, 316 Stainless Steel, Elbow, Compression Union, 1/8" OD
Representative image only.
Item # EW-31406-06
Get a secure seal at higher pressures
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
Free Certificate of Comliance
$25.50USD / Each
BUY MORE & SAVE
  • 1-9$25.50USD / Each
  • 10-49$22.95USD / Each
  • 50-99$21.68USD / Each
  • 100+$20.40USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting Material316 Stainless Steel
  • Sterilization MethodAutoclave, ethylene oxide (ETO), or gamma
  • Connection TypeCompression
  • Compression Size / Tube Od (Port 1) (in)1/8
  • Compression Size / Tube Od (Port 2) (in)1/8
  • DescriptionCompression Fitting, Union Elbow, 316 SS, 1/8" OD
Certificate of Compliance
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
More About this Item
Very good chemical resistance. Rated to 220 psi maximum at 77°F ( 25°C). Usable from 32 to 212°F (0 to 100°C).