Compression Fitting, Union Tee, 316 SS, 1/2" OD
Representative image only.
Item # EW-31406-20
$76.00USD / Each
BUY MORE & SAVE
  • 1-9$76.00USD / Each
  • 10-49$68.40USD / Each
  • 50-99$64.60USD / Each
  • 100+$60.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting Material316 Stainless Steel
  • Sterilization MethodAutoclave, ethylene oxide (ETO), or gamma
  • Connection TypeCompression
  • Compression Size / Tube Od (Port 1) (in)1/2
  • Compression Size / Tube Od (Port 2) (in)1/2
  • Compression Size / Tube Od (Port 3) (in)1/2
  • DescriptionCompression Fitting, Union Tee, 316 SS, 1/2" OD
Certificate of Compliance
More About this Item
Very good chemical resistance. Pressure limitation determined by the tubing, not the fitting. Usable from -40 to 200ºF (-40 to 93ºC).