Contact Plate, SabDex Agar, Friction Lid, for Surface Testing; 10/Pk
Representative image only.
Item # EW-14201-24
Cultivate microorganisms from environmental surfaces
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
PRODUCT OPTIONS
$43.00USD / Bag of 10
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)SabDex (Sabourand Dextrose) agar
  • DescriptionContact Plate, SabDex Agar, Friction Lid, for Surface Testing; 10/Pk
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
More About this Item
These high-quality, economical plates feature a specified grid molded into the bottom of the plate for easy colony counting.