Contact plate, TSA with Lecithin and Tween 80, 10/pk
Representative image only.
Item # EW-14201-20
Cultivate microorganisms from environmental surfaces
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
PRODUCT OPTIONS
$27.50USD / Bag of 10
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)Tryptic Soy w/ Lecithin, Tween
  • DescriptionContact plate, TSA with Lecithin and Tween 80, 10/pk
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
More About this Item
These high-quality, economical plates feature a specified grid molded into the bottom of the plate for easy colony counting.