Contec Prosat® PS-911EB Presaturated Cleanroom Wipes, Sterile; 30 Wipes/Pack, 12 Pks/Cs
Representative image only.
Contec Mfr # PS-911EB – Item # EW-33675-01
Sterile and prewetted Class 100 wipes
  • Features resealable pouch with a tamper evident label
$272.50USD / Case of 12
SPECS & DESCRIPTION
Specifications & Description
  • ComplianceClass 100
  • Width (in)11
  • Length (in)9
  • DescriptionPrewetted Sterile Wipes, 9\" x 11\", 30/pack, 12 packs/case
Key Features
  • Features resealable pouch with a tamper evident label
More About this Item
Prewetted Class 100 wipes are sterilized using an E-Beam irradiation process making them ideal for pharmaceutical and biotech applications. Wipes are constructed of nonwoven, meltblown polypropylene and come in a convenient resealable pouch with a tamper evident label. Each pouch contains 30 wipes. Case of 12 pouches.
Sterile wipes with 70% isopropyl alcohol/30% DI water solution. Measure 9" x 11"
Not available for sale to Canada or California