ABB RVDA6V40 Continental 1-Phase Solid-State Contactors; 660VAC/40A/DC Logic
Representative image only.
ABB Mfr # SSM1A445BD – Item # EW-94469-47
SPECS & DESCRIPTION
Specifications & Description
  • Load Rating660 VAC
  • Control Voltage4 to 32 VDC
  • Max Current (amps)40
  • Input LogicOn: 4 VDC (at 5.4 mA) to 32 VDC (at 10 mA)
  • DescriptionContinental 1-Phase Solid-State Contactors; 660VAC/40A/DC Logic
More About this Item
FEATURES
  • Superior Surge Survival Technology
  • 25 or 40 amp ratings
  • 575 or 660V max
  • Integrated heatsink
  • LED input indicator
  • Direct copper bonded SCRs
  • Mounts on DIN rail or panel
  • Meets En60947-4-3 and EN55011
  • UL recognized, CSA listed
  • Zero voltage turn-on (CIRVMA units)