CoorsTek 66128 Porcelain Crucible Covers, 90 mm ID; 6/Pk
Representative image only.
CoorsTek Mfr # 66128 – Item # EW-17811-14
  • Crucible covers are glazed, except for rim
  • Autoclavable
SPECS & DESCRIPTION
Specifications & Description
  • Cap MaterialPorcelain
  • AutoclavableYes
  • Inside Rim Diameter (mm)90
  • DescriptionPorcelain Crucible Covers, 90 mm ID; 6/Pk
Key Features
  • Crucible covers are glazed, except for rim
  • Autoclavable
More About this Item
Compatible cover for the following part numbers: 17803-16, 17803-36, 17804-12 and 17804-32