This item is now discontinued. Please browse similar items
CoorsTek 60281 Buchner Vacuum Funnels, Ceramic, 750 mL, 2/cs
Representative image only.
CoorsTek Mfr # 60281 – Item # EW-17817-22
SPECS & DESCRIPTION
Specifications & Description
  • MaterialCeramic
  • Capacity (mL)750
  • Overall Height (mm)109
  • DescriptionBuchner Vacuum Funnels, Ceramic, 750 mL, 2/cs
More About this Item
One piece tabletop filter with side outlet for vacuum filtration accepts standard filters. Funnels are glazed inside and out except for the rim. Tubulature OD is approximately 14.5 mm at widest point. Autoclavable.