CoorsTek 60319 Porcelain Mortar, 275 mL, pack of 1

Representative image only.
CoorsTek Mfr # 60319 – Item # EW-17820-06

$68.50USD / Pkg of 1

SPECS & DESCRIPTION

Specifications & Description

  • Mortars Capacity (mL)275
  • Mortars Top OD (mm)115
  • Mortars Height (mm)70
  • DescriptionPorcelain Mortar, 275 mL, pack of 1

More About this Item

Mortars are glazed all over except for grinding surfaces and the outside bottom. Features include lip and pour spout. Mortar and pestle are sold as separate pieces. Autoclavable.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat