CoorsTek 60374 99.5% Alumina Pestle, 117 mm Length; 1/Pk
Representative image only.
CoorsTek Mfr # 60374 – Item # EW-17823-30
$278.00USD / Case of 1
BUY MORE & SAVE
  • 1-4$278.00USD / Case of 1
  • 5-9$250.20USD / Case of 1
  • 10+$236.30USD / Case of 1
SPECS & DESCRIPTION
Specifications & Description
  • Pestles Length (mm)117
  • Description99.5% Alumina Pestle, 177 mm Length; 1/Pk
More About this Item
Ideal for precise elemental analysis where minimal sample contamination is required. Pestles are glazed all over except for grinding surfaces. Mortar and pestle are sold as separate pieces. Autoclavable.