CoorsTek 60437 Porcelain Spot Plates, Blue, 12-Well, 118mm L x 91mm W; 1/Pk

Representative image only.
CoorsTek Mfr # 66437 – Item # EW-17825-00

SPECS & DESCRIPTION

Specifications & Description

  • ColorBlue
  • MaterialPorcelain
  • Overall Length (in)4 5/8
  • Overall Length (cm)11.8
  • Overall Width (in)3 9/16
  • Overall Width (cm)9.1
  • DescriptionPorcelain Spot Plates, Blue, 12-Well, 118mm L x 91mm W; 1/Pk

More About this Item

Dark blue glaze provides a sharp contrast to easily observe white precipitates. Glazed all over except bottom. Autoclavable.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat