CoorsTek 66125 Porcelain Crucible Covers, 54 mm ID; 6/Pk
Representative image only.
CoorsTek Mfr # 66125 – Item # EW-17811-08
  • Crucible covers are glazed, except for rim
  • Autoclavable
$114.00USD / Pkg of 6
SPECS & DESCRIPTION
Specifications & Description
  • Cap MaterialPorcelain
  • AutoclavableYes
  • Inside Rim Diameter (mm)54
  • DescriptionPorcelain Crucible Covers, 54 mm ID; 6/Pk
Key Features
  • Crucible covers are glazed, except for rim
  • Autoclavable
More About this Item
Compatible cover for the following part numbers: 17803-10, 17803-12, 17803-30, 17803-32, 17804-06, 17804-26, 17805-06 and 17805-26.