CoorsTek 66323 Ceramic/Porcelain Pestle, 194 mm Length; 1/Pk
Representative image only.
CoorsTek Mfr # 66323 – Item # EW-17821-08
SPECS & DESCRIPTION
Specifications & Description
  • Pestles Length (mm)194
  • DescriptionCeramic/Porcelain Pestle, 194 mm Length; 1/Pk
More About this Item
Pestles are glazed all over except for grinding surfaces. Features an oversized pestle for comfort. Mortar and pestle are sold as separate pieces. Autoclavable.