Corning 3080-200LO Replacement bowl for 3080 200 mm desiccator
Representative image only.
Pyrex Mfr # 3080-200LO – Item # EW-34548-23
Large size, knob top
SPECS & DESCRIPTION
Specifications & Description
  • Chamber Volume (cu ft)0.26
  • Chamber Volume (Liters)7.5
  • DescriptionReplacement bowl for 3080 200 mm desiccator
More About this Item
These large desiccators are durable and efficient. Approximate bowl volume is 3800 mL on the 200-mm size and 7500 mL on the 250-mm size. Not for vaccum applications.
Do not heat or subject to pressure.