Corning 3366 96-Well Microplates, Immunology, PS, Round, High Binding
Representative image only.
Corning Mfr # 3366 – Item # EW-01728-22
Clear high-grade polystyrene for research diagnostic assays
  • Formulated for uniform binding, high optical clarity, and low background absorption
  • Medium-binding plates offer a binding capacity of 250 ng of mouse IgG/cm2
  • High-binding plates offer a binding capacity of 500 ng of mouse IgG/cm2
PRODUCT OPTIONS
$592.00USD / Case of 100
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (mL)0.33
  • Well Volume (µL)360
  • Well ShapeRound
  • SterileNo
  • Plate TypeSolid plates
  • Plate ColorClear
  • Binding PropertyHigh
  • MaterialPolystyrene
  • Description96-Well Microplates, Immunology, PS, Round, High Binding
Key Features
  • Formulated for uniform binding, high optical clarity, and low background absorption
  • Medium-binding plates offer a binding capacity of 250 ng of mouse IgG/cm2
  • High-binding plates offer a binding capacity of 500 ng of mouse IgG/cm2