Corning 3907 Roller Bottles, CellBind surface, 850 cm2
Representative image only.
Corning Mfr # 3907 – Item # EW-01835-40
Seamless, one-piece construction reduces risk of breakage
  • Cell culture treated for optimal cell attachment
  • Individually printed with lot numbers for easy identification and traceability
SPECS & DESCRIPTION
Specifications & Description
  • Descriptionroller bottles. CellBind surface, 850 cm2
Nonpyrogenic
Key Features
  • Cell culture treated for optimal cell attachment
  • Individually printed with lot numbers for easy identification and traceability
More About this Item
Optically-clear polystyrene roller bottles are sterilized by gamma radiation and certified nonpyrogenic. Bottles are tissue culture treated for optimal cell attachment. Bottles are constructed from durable polystyrene and caps are made of HDPE. Models 01836-05 and -06 feature a ribbed design that doubles the growth area without increasing bottle size. Two window areas provide microscopic viewing as well as pouring surfaces.
Choose from three cap type options. The plug seal provides a liquid-tight seal and the Easy grip cap is designed for easy opening. The Easy grip vent cap adds a 0.2 μm nonwetting (hydrophobic) membrane sealed to the cap.