Corning 3930 Polystyrene lids, Corner-notch, 100/cs
Representative image only.
Corning Mfr # 3930 – Item # EW-01730-01
Designed to reduce contamination
PRODUCT OPTIONS
$713.00USD / Case of 100
SPECS & DESCRIPTION
Specifications & Description
  • Plate TypeWell plate lid
  • DescriptionPolystyrene lids, Corner-notch, 100/cs
More About this Item
The clear polystyrene (PS) lids are well suited for the Corning 96-well plates. A corner-notch design with condensation rings reduces contamination. Available sterile.