Corning Roller Bottle, Sterile, Polystyrene, Tissue Culture Treated, Plug Seal, 490 cm2; 40/Cs
Representative image only.
Corning Mfr # 430195 – Item # EW-01836-00
Seamless, one-piece construction reduces risk of breakage
  • Cell culture treated for optimal cell attachment
  • Individually printed with lot numbers for easy identification and traceability
  • Bottles do not have graduations
  • 2 year warranty
PRODUCT OPTIONS
$440.00USD / Case of 40
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolystyrene Bottle and HDPE Cap
  • ClosurePlug Seal
  • Height (mm)173
  • Diameter (mm)111.25
  • Culture Area (cm²)490
  • SterileYes
  • Capacity (mL)150
  • DescriptionRoller Bottle, Sterile, Polystyrene, Tissue Culture Treated, Plug Seal, 490 cm2; 40/Cs
  • Warranty2 year
Two year warranty
Key Features
  • Cell culture treated for optimal cell attachment
  • Individually printed with lot numbers for easy identification and traceability
  • Bottles do not have graduations
  • 2 year warranty
More About this Item
Optically-clear polystyrene roller bottles are sterilized by gamma radiation and certified nonpyrogenic. Bottles are tissue culture treated for optimal cell attachment. Bottles are constructed from durable polystyrene and caps are made of HDPE. Models 01836-05 and -06 feature a ribbed design that doubles the growth area without increasing bottle size. Two window areas provide microscopic viewing as well as pouring surfaces.
Choose from three cap type options. The plug seal provides a liquid-tight seal and the Easy grip cap is designed for easy opening. The Easy grip vent cap adds a 0.2 μm nonwetting (hydrophobic) membrane sealed to the cap.
Plug seal caps feature one-piece linerless construction and are designed for use in closed systems, providing a liquid- and gas-tight seal. When loosened, this cap can also be used in open systems.