Corning Disposable Cap, 15-415 Thread, Phenolic with Rubber Liner; 1000/ Cs
Representative image only.
Corning Mfr # 99999-15 – Item # EW-34581-22
Keep your sample safe and secure
  • Constructed of black phenolic resin with white rubber liner
  • Autoclavable
$255.00USD / Case of 1000
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPhenolic and Rubber
  • ColorBlack
  • Compatible WithTubes with 15-415 Thread
  • Connections15-415
  • Height (mm)16
  • Diameter (mm)18
  • DescriptionDisposable Cap, 15-415 Thread, Phenolic with Rubber Liner; 1000/ Cs
Key Features
  • Constructed of black phenolic resin with white rubber liner
  • Autoclavable
More About this Item
These disposable Corning® caps are made from phenolic resin and are autoclavable at 121°C. Caps have GPI 15-415 threads and are provided with a white rubber liner.