Corning Gosselin BB64-01 Contact Petri Dish, 60 x 15mm, Sterile, 4 Vents, Domed Base; 720/Cs
Representative image only.
Corning Gosselin Mfr # BB64-01 – Item # EW-34551-79
Gridded and disposable contact dishes with a grip lid
  • Crystal clear for best readability
  • Keep agar at a constant depth—ultra-flat and even surface
  • Traceable records on every box
  • Reduce waste disposal costs—very light weight
PRODUCT OPTIONS
$411.00USD / Case of 720
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolystyrene
  • Closure4 vents
  • Depth (mm)65
  • Diameter (mm)145
  • SterileYes
  • DescriptionContact Petri Dish, 60 x 15mm, Sterile, 4 Vents, Domed Base; 720/Cs
Sterile
Includes
double bagged dishes with traceability labels.
Key Features
  • Crystal clear for best readability
  • Keep agar at a constant depth—ultra-flat and even surface
  • Traceable records on every box
  • Reduce waste disposal costs—very light weight
More About this Item
Crystal clear polystyrene (PS) dishes are heat resistant to 55°C and available sterile or aseptic. Dishes have a stacking ring which makes them practical to pile. All dishes have a grip lid and come with a flat or domed base. Pick vented dishes to reduce condensation and allow air circulation. 60 x 15 mm style dihes measure closer to 65 x 14.5 mm.
The packaging is 20 dishes to a sleeve placed in a sealed and traceable bag, 36 sleeves inside a second folded bag per case.