Corning Gosselin Sterile Petri Dish, Slippable, 100 x 15 mm, 25 per sleeve; 500/pk
Representative image only.
Corning Gosselin Mfr # BP94A-01 – Item # EW-34551-68
Culture dishes for any application
  • Injection point located near the edge of the bottom maintains optimal clarity for imaging systems Lightweight polystyrene (PS) is compliant with heavy metals (94/62/EC) and food contact (EC 1935/2004) directives Class 100 manufacturing environment ensures aseptic production and packaging Vented for easy gas exchange
PRODUCT OPTIONS
$100.00USD / Case of 500
SPECS & DESCRIPTION
Specifications & Description
  • MaterialOptically clear polystyrene
  • ClosureVented, slippable
  • Height (mm)15
  • Diameter (mm)100
  • SterileYes
  • DescriptionSterile Petri Dish, Slippable, 100 x 15 mm, 25 per sleeve; 500/pk
Key Features
  • Injection point located near the edge of the bottom maintains optimal clarity for imaging systems Lightweight polystyrene (PS) is compliant with heavy metals (94/62/EC) and food contact (EC 1935/2004) directives Class 100 manufacturing environment ensures aseptic production and packaging Vented for easy gas exchange
More About this Item
Standard petri dishes are available in nonsterile and sterile (SAL 10⁻⁶) versions, crystal or optically clear polystyrene, and feature economical packaging. Choose between automation-compatible, slippable, and stackable dishes.