Corning Gosselin Dipper with Removable Handle, Polypropylene, Sterile, Screw Cap, 125 mL; 100/CS
Representative image only.
Corning Gosselin Mfr # TAT52C-01 – Item # EW-34551-76
Removable handle makes sample collection quick and easy
  • Obtain excellent chemical resistance from all polypropylene (PP) construction
PRODUCT OPTIONS
$325.00USD / Case of 100
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene (PP)
  • Sample Volume125
  • Bottle TypeSterile Straight-Sided Jar with Screw Cap
  • Cap ConfigurationScrew
  • Height (cm)27.5
  • Diameter (cm)5.2
  • DescriptionDipper with Removable Handle, Polypropylene, Sterile, Screw Cap, 125 mL; 100/CS
Sterile
Key Features
  • Obtain excellent chemical resistance from all polypropylene (PP) construction
More About this Item
The Corning® Gosselin™ dippers are designed with a removable handle, making sample collection even easier. Simply take your sample, remove the handle, and send the shockproof container to your testing facility. Dippers are sterile and individually packed inside a traceable and tamper-evident zipped bag to avoid cross-contamination. Choose from bottles with a screw or hinge cap, ideal for any water sampling application.