Corning RND BOTTOM BOIL FLASK 500ML
Representative image only.
Pyrex Mfr # 4320B-500 – Item # EW-67972-59
Boiling, round bottom, short neck, joint
SPECS & DESCRIPTION
Specifications & Description
More About this Item
These boiling flasks with short neck are carefully designed and manufactured from borosilicate glass. Uniform wall thickness, a characteristic of all round-bottom, Pyrex glassware, provides a balance between expansion and mechanical strength for greater shock resistance and versatility. Use boiling flasks for a variety of mixing, heating, and boiling applications commonly encountered in most laboratory work. The normal working temperature is 230°>C. Includes a Ts joint.