Corning Stripette Serological Pipettes, Sterile, 2.0 mL; 200/Cs

Representative image only.
Corning Mfr # 4486 – Item # EW-13001-30
Bidirectional graduations provide choice of ascending and descending scales
  • Anti-drip tip assures accurate delivery
  • Color-coded magnifier stripes make volume reading and size selection easier
  • Negative graduations allow additional working volume
  • Calibrated "to deliver" for accurate dispensing to ±2%
  • Sterile, individually wrapped, nonpyrogenic, DNase-/RNase-free, and Human DNA-free

SPECS & DESCRIPTION

Specifications & Description

  • SterileYes
  • Max Volume (µL)2000
  • Min Volume (µL)0
  • Min Volume (mL)0
  • Max Volume (mL)2
  • Tip Volume (mL)2
  • Max Dispensing (mL)2
  • Graduations (mL)0.01
  • Accuracy (%)±2.00%
  • Overall Length (in)10.9
  • Overall Length (mm)277
  • Body MaterialPolystyrene
  • Color CodeGreen
  • Package OfIndividually wrapped
  • DescriptionSerological Pipettes, Sterile, 2.0 mL; 200/Cs

Key Features

  • Anti-drip tip assures accurate delivery
  • Color-coded magnifier stripes make volume reading and size selection easier
  • Negative graduations allow additional working volume
  • Calibrated "to deliver" for accurate dispensing to ±2%
  • Sterile, individually wrapped, nonpyrogenic, DNase-/RNase-free, and Human DNA-free

More About this Item

The Stripette™ serological pipettes are constructed of polystyrene and feature color-coded vertical stripes which are magnified by internal reagents, making the meniscus easy to read for quick volume identification. Uniform mouthpieces fit all standard pipette fillers. Individually wrapped paper/plastic packaging with "Banana peel" design make pipettes easy to open and remove aseptically with one hand, ideal for sterile tissue culture applications, especially while wearing latex gloves, as the wrap reduces static cling.

Uniform mouthpieces fit all standard pipette fillers. "Banana peel" (paper/plastic-wrap) packaging make them easy to open and remove pipettes aseptically with one hand.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat