Costar 3960 96-Well Assay Blocks, Nontreated, PP, 2.0 mL, Sterile
Representative image only.
Costar Mfr # 3960 – Item # EW-01729-03
96-Well Microplates and Assay Blocks, Nontreated, PP
  • Constructed of clarified polypropylene for optimal clarity and solvent resistance—ideal for compound library and combinatorial chemistry applications
PRODUCT OPTIONS
$303.00USD / Case of 25
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (mL)2
  • Well ShapeV-bottom
  • Surface TreatmentUntreated
  • SterileYes
  • Plate TypeDeep-well assay block
  • MaterialPolypropylene
  • Description96-Well Assay Blocks, Nontreated, PP, 2.0 mL, Sterile
Key Features
  • Constructed of clarified polypropylene for optimal clarity and solvent resistance—ideal for compound library and combinatorial chemistry applications
More About this Item
Standard microplates are built to industry standard footprint; compatible with automated systems. Plates are certified RNase- and Dnase-free.