CPC (Colder) AseptiQuik G® Quick-Disconnect Fitting, Genderless Hose Barb Connector, Polycarbonate, 1/4" ID; 5/Pk
Representative image only.
CPC (Colder) Mfr # AQG17004 – Item # EW-20202-40
Provide easy, secure sterile connections even in nonsterile environments
  • Genderless design simplifies system integration—no need to worry about female and male threads
  • Robust construction provides reliable performance—even without clamps
  • Quick, three-step connection process reduces risk of operator error
  • Quick-disconnect fitting features 1/4" ID hose barb connection
PRODUCT OPTIONS
$205.00USD / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolycarbonate
  • Seal MaterialPlatinim-cured silicone
  • Sterilization MethodGamma
  • Connection TypeQuick-Disconnect x Barb
  • Barb Size / Tubing Id (Port 1) (in)1/4
  • ColorWhite with Blue Tab Covers
  • Min Temperature (° F)34
  • Max Temperature (° F)104
  • Min Temperature (° C)1
  • Max Temperature (° C)40
  • Max Operating Pressure (PSI)75
  • DescriptionAseptiQuik-G® Quick-Disconnect Fitting, Hose Barb Connector, Polycarbonate, 1/4" ID; 5/Pk
Key Features
  • Genderless design simplifies system integration—no need to worry about female and male threads
  • Robust construction provides reliable performance—even without clamps
  • Quick, three-step connection process reduces risk of operator error
  • Quick-disconnect fitting features 1/4" ID hose barb connection
More About this Item
Manufactured from sturdy polycarbonate, these genderless connectors are ideal for single-use and sanitary applications. Use hose barb connectors with filters, tubing, and process containers to create systems for upstream or downstream processes.
Simply fit the connector half in your tubing line. The easy three-step FLIP-CLICK-PULL assembly procedure ensures a tight seal with no additional hardware required. The audible “click” confirms a securely connected assembly.
All fittings can be sterilized by gamma irradiation. Use high-temperature fittings to sterilize via autoclave.