CPC (Colder) AseptiQuik-S® Quick-Disconnect Fitting, Sanitary Connector, Polycarbonate, High Temperature, 3/4" Mini Tri-Clamp; 1/Each
Representative image only.
CPC (Colder) Mfr # AQS33012HT – Item # EW-20002-96
Provide easy, secure, sterile connections even in nonsterile environments
  • Quick, three-step connection process reduces risk of operator error
  • Large housing size allows easy installation
  • Quick-disconnect fitting features 3/4" Mini Tri-Clamp connection
  • High temperature version allows for gamma and autoclave sterilization
PRODUCT OPTIONS
$47.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolycarbonate
  • Seal MaterialPlatinim-cured silicone
  • Sterilization MethodAutoclave or gamma
  • Connection TypeQuick-Disconnect x Sanitary
  • Sanitary Clamp Size (Port 1) (in)3/4
  • ColorWhite with White Tab Covers
  • Min Temperature (° F)39
  • Max Temperature (° F)104
  • Min Temperature (° C)4
  • Max Temperature (° C)40
  • Max Operating Pressure (PSI)60
  • DescriptionAseptiQuik-S® Quick-Disconnect Fitting, Sanitary Connector, Polycarbonate, High Temperature, 3/4" Mini Tri-Clamp; 1/Each
Key Features
  • Quick, three-step connection process reduces risk of operator error
  • Large housing size allows easy installation
  • Quick-disconnect fitting features 3/4" Mini Tri-Clamp connection
  • High temperature version allows for gamma and autoclave sterilization
More About this Item
Manufactured from sturdy polycarbonate, these genderless connectors are ideal for single-use and sanitary applications. Use connectors with filters, tubing, and process containers to create systems for upstream or downstream processes.
Simply fit the connector half in your tubing line. Push the connectors together; an audible snap confirms a tight connection. Align and snap membrane connectors together and twist blue lock ring clockwise to hear another audible click. Alignment of the ring rib with the body’s arrow indicator confirms final, secure connection.
All fittings can be sterilized by gamma irradiation. Use high temperature fittings to sterilize via autoclave.