This item is now discontinued. Please browse similar items
CPC (Colder) HFC12812 Coupling Body, Panel Mount, Polypropylene (PP), 1/2" OD Tube; 10/PK
Representative image only.
CPC (Colder) Mfr # HFC12812 – Item # EW-31308-50
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Seal MaterialEPDM
  • Sterilization MethodGamma or e-beam
  • Connection TypeQuick-Disconnect x Compression
  • Compression Size / Tube Od (Port 1) (in)1/2
  • Quick-Disconnect Flow Size (in)3/8
  • Min Temperature (° F)32
  • Max Temperature (° F)160
  • Min Temperature (° C)0
  • Max Temperature (° C)71
  • Max Operating Pressure (PSI)60
  • Fitting Length (in)57/64
  • DescriptionCoupling Body, Panel Mount, 1/2" OD Tube
More About this Item
High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately). Plug insert into coupling body to connect—fitting halves lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Designed to enable enhanced flow in a coupling with a 3/8" flow path, the HFC series is available in polypropylene and polysulfone for a wide variety of applications. The HFC series has an ergonomic, easy-to-operate design and comes standard with a shrouded latch to protect against accidental disconnect. UV-resistant polysulfone Couplings of UV-resistant polysulfone and with BSP threaded terminations are also available; contact Cole-Parmer for more information.