This item is now discontinued. Please browse similar items
CPC (Colder) HFC161235 Polysulfone straight-through bulkhead panel mount hose barb, 3/8" flow size, 3/4" ID; 10/PK
Representative image only.
CPC (Colder) Mfr # HFC161235 – Item # EW-31308-71
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolysulfone
  • Sterilization MethodAutoclave, ethylene oxide (ETO), or gamma
  • Connection TypeQuick-Disconnect x Barb
  • Barb Size / Tubing Id (Port 1) (in)3/4
  • Quick-Disconnect Flow Size (in)3/8
  • Min Temperature (° F)-40
  • Max Temperature (° F)280
  • Min Temperature (° C)-40
  • Max Temperature (° C)137
  • Max Operating Pressure (PSI)125
  • Max Vacuum (in Hg)27.5
  • Max Vacuum (mm Hg)698
  • DescriptionPolysulfone straight-through bulkhead panel mount hose barb, 3/8" flow size, 3/4" ID
More About this Item
High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately). Plug insert into coupling body to connect—fitting halves lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 SS valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 SS latch springs and EPDM seals.