CPC (Colder) HFC22839M High-Flow Coupling Insert, Straight-Through, Polysulfone, 1/2" Barbed Connection
Representative image only.
CPC (Colder) Mfr # HFC22839M – Item # EW-31313-13
$114.00USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$114.00USD / Pkg of 10
  • 10-49$102.60USD / Pkg of 10
  • 50-99$96.90USD / Pkg of 10
  • 100+$91.20USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolysulfone
  • Sterilization MethodAutoclave or gamma
  • Connection TypeQuick-Disconnect x Barb
  • Barb Size / Tubing Id (Port 1) (in)1/2
  • DescriptionHigh-flow coupling insert, straight-through, polysulfone, 1/2" barbed connection
More About this Item
USP Class VI-compliant couplings require a fitting body and an insert (order separately). Fitting materials are BSE/TSE-free. Valves restrict flow when fitting halves are disconnected.