CPC (Colder) HFCD22812 Quick-disconnect fittings, Valved hose barbs inserts, PP, 1/2"
Representative image only.
CPC (Colder) Mfr # HFCD22812 – Item # EW-31303-46
$14.80USD / Each
BUY MORE & SAVE
  • 1-9$14.80USD / Each
  • 10-49$13.32USD / Each
  • 50-99$12.58USD / Each
  • 100+$11.84USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Sterilization MethodEthylene oxide (ETO) or gamma
  • Connection TypeQuick-Disconnect x Barb
  • Barb Size / Tubing Id (Port 1) (in)1/2
  • Quick-Disconnect Flow Size (in)3/8
  • Min Temperature (° F)32
  • Max Temperature (° F)160
  • Min Temperature (° C)0
  • Max Temperature (° C)71
  • Max Operating Pressure (PSI)60
  • Max Vacuum (in Hg)27.5
  • Max Vacuum (mm Hg)698
  • DescriptionQuick-disconnect fittings, Valved hose barbs inserts, PP, 1/2"
More About this Item
High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately). Plug insert into coupling body to connect—fitting halves lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 SS valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 SS latch springs and EPDM seals.