CPC (Colder) High-Flow Quick-Disconnect Fitting, Threaded Insert, Polysulfone, Straight-Through, 1/2 NPT(M)
Representative image only.
CPC (Colder) Mfr # HFC24835 – Item # EW-31307-31
No more leaks—valved couplings automatically shut off flow
  • Compact, lightweight couplings feature a high flow and low spillage design
  • Not medical-grade Polysulfone
PRODUCT OPTIONS
$11.90USD / Each
BUY MORE & SAVE
  • 1-9$11.90USD / Each
  • 10-49$10.71USD / Each
  • 50-99$10.12USD / Each
  • 100+$9.52USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolysulfone
  • Seal MaterialEPDM
  • Valve Spring Material316 Stainless Steel
  • Sterilization MethodAutoclave, ethylene oxide (ETO), or gamma
  • Connection TypeQuick-Disconnect x NPT(M)
  • Thread size NPT(M) (port 1) (in)1/2
  • Quick-Disconnect Flow Size (in)3/8
  • Min Temperature (° F)-40
  • Max Temperature (° F)280
  • Min Temperature (° C)-40
  • Max Temperature (° C)137
  • Max Operating Pressure (PSI)125
  • Max Vacuum (in Hg)27.5
  • Max Vacuum (mm Hg)698
  • DescriptionHigh-Flow Quick-Disconnect Fitting, Threaded Insert, Polysulfone, Straight-Through, 1/2 NPT(M); 1/Ea
Key Features
  • Compact, lightweight couplings feature a high flow and low spillage design
  • Not medical-grade Polysulfone
More About this Item
These CPC® high-flow quick-disconnect fittings have flow comparable to many 1/2" flow couplings in a 3/8” body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 stainless steel valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 stainless steel latch springs and EPDM seals.
For a complete coupling, order one fitting body and one insert.