CPC (Colder) LC23006 Metal Quick-Disconnect Hose Barb Elbow Insert; Straight-through, 3/8" Tube ID; 10/PK
Representative image only.
CPC (Colder) Mfr # LC23006 – Item # EW-31309-02
$190.00USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialBrass
  • Seal MaterialBuna N
  • Sterilization MethodAutoclave or ethylene oxide (ETO)
  • Connection TypeQuick-Disconnect x Barb
  • Barb Size / Tubing Id (Port 1) (in)3/8
  • Quick-Disconnect Flow Size (in)1/4
  • Min Temperature (° F)-40
  • Max Temperature (° F)180
  • Min Temperature (° C)-40
  • Max Temperature (° C)82
  • Max Operating Pressure (PSI)250
  • Length (cm)3.2512
  • DescriptionMetal Quick-Disconnect Hose Barb Elbow Insert; Straight-through, 3/8" Tube ID
More About this Item
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180°F, and can be used with gases or liquids.