CPC (Colder) MC20025 Metal Quick-Disconnect Compression Insert; Straight-through, 5/32" Tube OD; 10/PK
Representative image only.
CPC (Colder) Mfr # MC20025 – Item # EW-31309-11
$154.00USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialBrass
  • Seal MaterialBuna N
  • Sterilization MethodAutoclave or ethylene oxide (ETO)
  • Connection TypeQuick-Disconnect x Compression
  • Compression Size / Tube Od (Port 1) (in)5/32
  • Quick-Disconnect Flow Size (in)1/8
  • Max Temperature (° F)180
  • Max Temperature (° C)82
  • Max Operating Pressure (PSI)250
  • Length (cm)2.8702
  • DescriptionMetal Quick-Disconnect Compression Insert; Straight-through, 5/32" Tube OD
More About this Item
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180°F, and can be used with gases or liquids.