CPC (Colder) Metal Quick-Disconnect Fitting, NPT (M) Threaded Insert, Valved, 1/4" NPT(M)
Representative image only.
CPC (Colder) Mfr # MCD2404 – Item # EW-31404-61
Convenience of a quick-disconnect in pressure and vacuum applications
  • Constructed of durable chrome-plated brass
  • Easy one-hand connection and disconnection
PRODUCT OPTIONS
$19.10USD / Each
BUY MORE & SAVE
  • 1-9$19.10USD / Each
  • 10-49$17.19USD / Each
  • 50-99$16.24USD / Each
  • 100+$15.28USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialChrome-Plated Brass
  • Seal MaterialBuna N
  • Valve Spring Material316 Stainless Steel
  • Sterilization MethodAutoclave or ethylene oxide (ETO)
  • Connection TypeQuick-Disconnect x NPT(M)
  • Thread size NPT(M) (port 1) (in)1/4
  • Quick-Disconnect Flow Size (in)1/8
  • Flow PatternValved
  • Min Temperature (° F)-40
  • Max Temperature (° F)180
  • Min Temperature (° C)-40
  • Max Temperature (° C)82
  • Max Operating Pressure (PSI)250
  • Length (cm)3.302
  • DescriptionMetal Quick-Disconnect Fittings, NPT (M) Threaded Insert, Valved, 1/4" NPT(M); 1/Ea
Key Features
  • Constructed of durable chrome-plated brass
  • Easy one-hand connection and disconnection
More About this Item
These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from -40 to 180°F, and can be used with gases or liquids.
A complete coupler requires a fitting body and an insert with the same flow size (order separately).