CPC (Colder) MPC™ Quick-Disconnect Fitting, Sanitary Insert, Polysulfone, Non-Valved, 3/4" Sanitary; 1/Ea

Representative image only.
CPC (Colder) Mfr # MPC44012T39M – Item # EW-50112-83
Connect to a coupling body to quickly create a sterile fluid pathway
  • Connect to a female coupling body for easy media transfer
  • Mix and match termination sizes for added flexibility
  • 0.375" nominal flow path size
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Compatible with MPC series body
  • 3/4" sanitary connection

PRODUCT OPTIONS

$24.00USD / Each
Connect a coupling body for a complete quick disconnect fitting

SPECS & DESCRIPTION

Specifications & Description

  • Fitting MaterialPolysulfone
  • O-Ring MaterialSilicone
  • Sterilization MethodGamma irradiation up to 50 kGy or autoclave up to 132°C (270°F) for 60 minutes, up to 25 repetitions
  • Connection TypeQuick Disconnect Body x Sanitary
  • Sanitary Clamp Size (Port 1) (in)3/4
  • Flow PatternStraight
  • ColorAmber tint
  • Min Temperature (° F)-40
  • Max Temperature (° F)300
  • Min Temperature (° C)-40
  • Max Temperature (° C)149
  • Max Operating Pressure (PSI)60
  • Fitting Length (in)1 13/32
  • Fitting Width (in)1
  • Fitting Length (mm)35.6
  • Fitting Width (mm)24.9
  • Regulatory ApprovalsNon-Phthalate;ADCF;USP;FDA;Non-DEHP;RoHS
  • DescriptionQuick-Disconnect Fitting, Sanitary Insert, Polysulfone, Non-Valved, 3/4\" Sanitary; 1/Ea

Key Features

  • Connect to a female coupling body for easy media transfer
  • Mix and match termination sizes for added flexibility
  • 0.375" nominal flow path size
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Compatible with MPC series body
  • 3/4" sanitary connection

More About this Item

The CPC® (Colder) MPC™/MPX® series connectors provide a simple way to transfer media between single-use systems. Connect a male insert and a female body to create a quick and secure fluid flow. Simply use a cap to seal off the fluid pathway. Coupling halves can rotate when connected reducing tube kinks. Select from various termination size options to suit your application.

All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat